Anti-ATG9A Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-4010R-CY5.5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Autophagy Related 9A
- Antigen symbol:ATG9A
- Clonality:Polyclonal
- Conjugation:Cy5.5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:79065
- Antigen synonyms:APG9L1|ATG9A|MGD3208|APG9-like 1|mATG9
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:321-371/839
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Involved in autophagy and cytoplasm to vacuole transport (Cvt) vesicle formation. Plays a key role in the organization of the preautophagosomal structure/phagophore assembly site (PAS), the nucleating site for formation of the sequestering vesicle. Cycles between a juxta-nuclear trans-Golgi network compartment and late endosomes. Nutrient starvation induces accumulation on autophagosomes. Starvation-dependent trafficking requires ULK1, ATG13 and SUPT20H.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ATG9A
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat