Order Entry
Norway
ContactUsLinkComponent
Anti-BCL2L11 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anti-BCL2L11 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-3058R-A647
Supplier:  Bioss
Anti-BCL2L11 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-3058R-A647
Supplier:  Bioss

Specifications

  • Antistofftype:
    Primary
  • Antigen-navn:
    BCL2-Like 11
  • Antigensymbol:
    BCL2L11
  • Klonalitet:
    Polyclonal
  • Konjugering:
    Alexa Fluor® 647
  • Vert:
    Rabbit
  • ImmunoKjemi:
    Yes
  • Isotype:
    IgG
  • Reaktivitet:
    Human,
    Rat,
    Mouse
  • Kyssabsorpsjon:
    No
  • Skjema:
    liquid
  • Gen-ID:
    12125
  • Antigen synonymer:
    BAM|BIM|Bcl2-L-11|Bcl-2-like protein 11|Bim|BOD|Bcl2-interacting mediator of cell death
  • Modifikasjon:
    phospho-Ser55
  • Lagringsbuffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Lagringstemp:
    Store at 4°C for 12 months
  • Konsentrasjon:
    1 μg/μl
  • Frakttemp.:
    4°C
  • Immunogen:
    30-70/196
  • Rensing:
    Purified by Protein A
  • Fp:
    100 µl

Specifications

About this item

Induces apoptosis and anoikis. The isoforms vary in cytotoxicity with isoform BimS being the most potent and isoform BimEL being the least potent.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BCL2L11
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat