Anti-ADAMTS5 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
BOSSBS-3573R-A555
: Bioss
- Antibody type:Primary
- Antigen name:ADAM Metallopeptidase with Thrombospondin Type 1 Motif 5
- Antigen symbol:ADAMTS5
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:11096
- Antigen synonyms:ADAMTS-11|ADAM-TS 5|ADAMTS-5|ADAMTS11|ADAM-TS5|ADAM-TS 11|ADMP-2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:530-580/930
- Purification:Purified by Protein A
- Pk:100 µl
Cleaves aggrecan, a cartilage proteoglycan, and may be involved in its turnover. May play an important role in the destruction of aggrecan in arthritic diseases. May play a role in proteolytic processing mostly during the peri-implantation period.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ADAMTS5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat