Anti-ND5 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
BOSSBS-3952R-A647
: Bioss
- Antibody type:Primary
- Antigen name:NADH dehydrogenase 5
- Antigen symbol:ND5
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Gene ID:4540
- Antigen synonyms:ND5|MTND5|NADH dehydrogenase subunit 5|NADH-ubiquinone oxidoreductase chain 5|NADH5|MT-ND5
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:502-552/603
- Purification:Purified by Protein A
- Pk:100 µl
Core subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I) that is believed to belong to the minimal assembly required for catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone (By similarity).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ND5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human