Order Entry
Norway
ContactUsLinkComponent
Anti-SIRT1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anti-SIRT1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-3393R-A488
Leverandør:  Bioss
Anti-SIRT1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-3393R-A488
Leverandør:  Bioss

Spesifikasjoner

  • Antibody type:
    Primary
  • Antigen name:
    Sirtuin 1
  • Antigen symbol:
    SIRT1
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 488
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    23411
  • Antigen synonyms:
    NAD-dependent Deacetylase Sirtuin-1|SIR2L1|SIRT1|SIR2-like Protein 1
  • Modification:
    phospho-Ser47
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    20-60/747
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Spesifikasjoner

Om dette produktet

This gene encodes a member of the sirtuin family of proteins, homologs to the yeast Sir2 protein. Members of the sirtuin family are characterized by a sirtuin core domain and grouped into four classes. The functions of human sirtuins have not yet been determined; however, yeast sirtuin proteins are known to regulate epigenetic gene silencing and suppress recombination of rDNA. Studies suggest that the human sirtuins may function as intracellular regulatory proteins with mono-ADP-ribosyltransferase activity. The protein encoded by this gene is included in class I of the sirtuin family. Alternative splicing results in multiple transcript variants.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SIRT1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse