Anti-KLK3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-3003R-A647
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Kallikrein-related peptidase 3
- Antigen symbol:KLK3
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:354
- Antigen synonyms:PSA|APS|hK3|KLK2A1
- Modification:phospho-Ser597
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
APS (adapter protein with Pleckstrin homology and Src homology 2 domains) is a member of the Lnk family, an adaptor protein that is involved in B cell signaling, insulin signaling and cytoskeletal reorganisation. A PH and an SH2 domain containing adaptor protein that links activated tyrosine kinases to signaling pathways. It is tyrosine phosphorylated by JAK2, KIT and other kinases during B cell receptor stimulation of many different cytokines, chemokines and leukokines. APS has been shown to inhibit the JAK STAT pathway in collaboration with cCbl.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: KLK3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat