Specifications
- Antibody type:Primary
- Antigen name:Ribosomal protein S6 kinase beta-1
- Antigen symbol:S6K1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:p70-S6K 1|alpha 1|S6K-beta-1|alpha 2|70 kDa ribosomal protein S6 kinase 1|p70 S6 kinase|P70S6K1|p70 S6K-alpha|serine/threonine-protein kinase 14A|ribosomal protein S6 kinase beta-1|p70 S6 kinase alpha|p70 ribosomal S6 kinase alpha|serine/threonine kinase 14 alpha|p70 S6KA|ribosomal protein S6 kinase I
- Modification:phospho-Thr444/Ser447
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
This gene encodes a member of the ribosomal S6 kinase family of serine/threonine kinases. The encoded protein responds to mTOR (mammalian target of rapamycin) signaling to promote protein synthesis, cell growth, and cell proliferation. Activity of this gene has been associated with human cancer. Alternatively spliced transcript variants have been observed. The use of alternative translation start sites results in isoforms with longer or shorter N-termini which may differ in their subcellular localizations. There are two pseudogenes for this gene on chromosome 17. [provided by RefSeq, Jan 2013].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: S6K1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat