Order Entry
Norway
ContactUsLinkComponent
 
Anti-MUC13 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
  BOSSBS-10074R-A555
 :  Bioss
Anti-MUC13 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
  BOSSBS-10074R-A555
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Mucin 13 Cell Surface Associated
  • Antigen symbol:
    MUC13
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 555
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    56667
  • Antigen synonyms:
    cell surface associated|MUC-13|MUC13 Down regulated in colon cancer 1|MUC13_HUMAN.|RECC|DRCC1|UNQ6194|Mucin 13|MUC 13
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

MUC13 is a mucin. These are secreted or transmembrane glycoproteins that are expressed mainly in the digestive tract and on epithelial surfaces. MUC13 is upregulated in gastric cancer. Mucins have also been implicated in inflammatory bowel diseases.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: MUC13
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat