Anti-BCL2L11 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
BOSSBS-3058R-HRP
: Bioss
- Antibody type:Primary
- Antigen name:BCL2-Like 11
- Antigen symbol:BCL2L11
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Gene ID:12125
- Antigen synonyms:BAM|BIM|Bcl2-L-11|Bcl-2-like protein 11|Bim|BOD|Bcl2-interacting mediator of cell death
- Modification:phospho-Ser55
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:30-70/196
- Purification:Purified by Protein A
- Pk:100 µl
Induces apoptosis and anoikis. The isoforms vary in cytotoxicity with isoform BimS being the most potent and isoform BimEL being the least potent.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: BCL2L11
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat