Anti-PDK2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-3962R-A350
Leverandør: Bioss
Spesifikasjoner
- Antibody type:Primary
- Antigen name:Pyruvate Dehydrogenase Kinase 2
- Antigen symbol:PDK2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:5164
- Antigen synonyms:Pyruvate dehydrogenase kinase isoform 2|pyruvate dehydrogenase kinase|PDK2_HUMAN.|mitochondrial|PDHK2|Pyruvate dehydrogenase lipoamide kinase isozyme 2|isozyme 2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Spesifikasjoner
Om dette produktet
This gene encodes a member of the pyruvate dehydrogenase kinase family. The encoded protein phosphorylates pyruvate dehydrogenase, down-regulating the activity of the mitochondrial pyruvate dehydrogenase complex. Overexpression of this gene may play a role in both cancer and diabetes. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene. [provided by RefSeq, Dec 2010].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PDK2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat