Anti-NEUROG3 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-0922R-CY5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Neurogenin 3
- Antigen symbol:NEUROG3
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:50674
- Antigen synonyms:NGN|bHLHa7|Atoh5|NGN1|Neurogenic basic-helix-loop-helix- protein|NGN-3|ngn3|Neurogenic differentiation factor 3|NEUROG1|NeuroD3|NEUROD3|Math4B
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:60-110/214
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Acts as a transcriptional regulator. Together with NKX2-2, initiates transcriptional activation of NEUROD1. Involved in neurogenesis. Also required for the specification of a common precursor of the 4 pancreatic endocrine cell types (By similarity).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Neurogenin 3
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat