Anti-NFKBIB Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-10075R-CY7
Leverandør: Bioss
Spesifikasjoner
- Antibody type:Primary
- Antigen name:Nuclear Factor Of Kappa Light Polypeptide Gene Enhancer In B-Cells Inhibitor, Beta
- Antigen symbol:NFKBIB
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:4793
- Antigen synonyms:IkBb|ikB-B|IKappaBbeta|I-kappa-B-beta|I(Kappa)B(beta)|IKB-beta|NF-kappa-BIB|IkB
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Spesifikasjoner
Om dette produktet
Nidogen 2 is an adhesion glycoprotein which is widely distributed in basement membranes. Binds to collagens I and IV, to perlecan and to laminin 1. Does not bind fibulins. It probably has a role in cell extracellular matrix interactions.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Nidogen2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat