Anti-MTF1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
BOSSBS-3601R-FITC
: Bioss
- Antibody type:Primary
- Antigen name:Metal-regulatory Transcription Factor 1
- Antigen symbol:MTF1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:4520
- Antigen synonyms:MRE-binding transcription factor|MTF1|MTF-1|ZRF|Transcription factor MTF-1|Metal regulatory transcription factor 1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:110-160/753
- Purification:Purified by Protein A
- Pk:100 µl
Activates the metallothionein I promoter. Binds to the metal responsive element (MRE).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: MTF1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat