Order Entry
Norway
ContactUsLinkComponent
 
Anti-ADRB2 Rabbit Polyclonal Antibody (Cy5.5®)
  BOSSBS-0947R-CY5.5
 :  Bioss
Anti-ADRB2 Rabbit Polyclonal Antibody (Cy5.5®)
  BOSSBS-0947R-CY5.5
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Adrenoceptor Beta 2
  • Antigen symbol:
    ADRB2
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy5.5®
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    154
  • Antigen synonyms:
    BETA2AR|ADRBR|Beta-2 adrenoreceptor|BAR|ADRB2R|Beta-2 adrenoceptor|B2AR
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    221-274/413
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

Beta-adrenergic receptors mediate the catecholamine-induced activation of adenylate cyclase through the action of G proteins. The beta-2-adrenergic receptor binds epinephrine with an approximately 3-fold greater affinity than it does norepinephrine.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADRB2
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat