Specifications
- Antibody type:Primary
- Antigen symbol:HP1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Multiple
- Cross adsorption:No
- Form:liquid
- Gene ID:34119
- Modification:phospho-Tyr43
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
HP1 alpha is a component of heterochromatin. It recognizes and binds histone H3 tails methylated at Lys-9, leading to epigenetic repression. HP1 alpha may interact with lamin B receptor (LBR). This interaction can contribute to the association of the heterochromatin with the inner nuclear membrane. HP1 proteins are relatively small proteins (~25 kDa) with a conserved amino-terminal chromo domain and a structurally related carboxy-terminal motif, the chromo shadow domain. Both domains of HP1 are required for binding to native chromatin in vivo, but they contribute differentially to binding in euchromatin and heterochromatin.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HP1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Other Species