Anti-ACTC1 Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-10196R-CY3
Leverandør: Bioss
Spesifikasjoner
- Antibody type:Primary
- Antigen name:Actin, Alpha, Cardiac Muscle 1
- Antigen symbol:ACTC1
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:70
- Antigen synonyms:CMD1R|LVNC4|CMH11|ACTC|ASD5
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:190-240/377
- Purification:Purified by Protein A
- Pk:100 µl
Spesifikasjoner
Om dette produktet
Actins are highly conserved proteins that are involved in various types of cell motility and are ubiquitously expressed in all eukaryotic cells.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ACTC1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat