Order Entry
Norway
ContactUsLinkComponent
 
Anti-CLEC5A Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  BOSSBS-2663R-FITC
 :  Bioss
Anti-CLEC5A Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  BOSSBS-2663R-FITC
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    C-type lectin Domain family 5, member a
  • Antigen symbol:
    CLEC5A
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    23845
  • Antigen synonyms:
    Mdl1|Clecsf5|MDL-1|Ly100
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    90-130/190
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

Functions as a positive regulator of osteoclastogenesis. Cell surface receptor that signals via TYROBP. Regulates inflammatory responses. Acts as a key regulator of synovial injury and bone erosion during autoimmune joint inflammation. Critical macrophage receptor for dengue virus serotypes 1-4. The binding of dengue virus to CLEC5A triggers signaling through phosphylation of TYROBP, this interaction does not result in viral entry but stimulates proinflammatory cytokine release.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CLEC5A
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype:
Reactivity: