Anti-IL21R Rabbit Polyclonal Antibody (Cy5.5®)
BOSSBS-2622R-CY5.5
: Bioss
- Antibody type:Primary
- Antigen name:Interleukin 21 Receptor
- Antigen symbol:IL21R
- Clonality:Polyclonal
- Conjugation:Cy5.5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:50615
- Antigen synonyms:CD360|IL-21 Receptor|Novel interleukin receptor|UNQ3121/PRO10273|Interleukin-21 receptor|IL-21 receptor|NILR
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:470-538/538
- Purification:Purified by Protein A
- Pk:100 µl
This is a receptor for interleukin-21.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: IL21R
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat