Anti-KIR2DL4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BOSSBS-2643R-HRP
Leverandør: Bioss
Spesifikasjoner
- Antibody type:Primary
- Antigen name:Killer cell immunoglobulin-like Receptor two Domains long cytoplasmic tail 4
- Antigen symbol:KIR2DL4
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Gene ID:3805
- Antigen synonyms:KIR103|killer cell immunoglobulin-like receptor 2DL4|natural killer cell inhibitory receptor|killer Ig receptor|killer cell inhibitory receptor 103AS|MHC class I NK cell receptor KIR103AS|CD158 antigen-like family member D|NK cell receptor|KIR103AS|CD158D|G9P|KIR-103AS
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:40-90/377
- Purification:Purified by Protein A
- Pk:100 µl
Spesifikasjoner
Om dette produktet
Receptor on natural killer (NK) cells for HLA-C alleles. Inhibits the activity of NK cells thus preventing cell lysis.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: KIR2DL4
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human