Anti-EEF1A1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-2503R-A350
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Eukaryotic Translation Elongation Factor 1 Alpha 1
- Antigen symbol:EEF1A1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:1915
- Antigen synonyms:EEF-1|EF1A|EE1A1|GRAF-1EF|CCS-3|EF-Tu|LENG7|PTI1|eEF1A-1|HNGC:16303|EEF1A|CCS3
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:145-178/462
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
This protein promotes the GTP-dependent binding of aminoacyl-tRNA to the A-site of ribosomes during protein biosynthesis. With PARP1 and TXK, forms a complex that acts as a T helper 1 (Th1) cell-specific transcription factor and binds the promoter of IFN-gamma to directly regulate its transcription, and is thus involved importantly in Th1 cytokine production.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: EEF1A1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat