Order Entry
Norway
ContactUsLinkComponent
Anti-CCL19 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anti-CCL19 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-2454R-A555
Supplier:  Bioss
Anti-CCL19 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-2454R-A555
Supplier:  Bioss

Specifications

  • Antistofftype:
    Primary
  • Antigen-navn:
    Chemokine (C-C motif) ligand 19
  • Antigensymbol:
    CCL19
  • Klonalitet:
    Polyclonal
  • Konjugering:
    Alexa Fluor® 555
  • Vert:
    Rabbit
  • ImmunoKjemi:
    Yes
  • Isotype:
    IgG
  • Reaktivitet:
    Human,
    Rat,
    Mouse
  • Kyssabsorpsjon:
    No
  • Skjema:
    liquid
  • Gen-ID:
    24047
  • Antigen synonymer:
    MIP3B|Scya19|ELC|exodus-3|CKb11
  • Lagringsbuffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Lagringstemp:
    Store at 4°C for 12 months
  • Konsentrasjon:
    1 μg/μl
  • Frakttemp.:
    4°C
  • Immunogen:
    60-100/108
  • Rensing:
    Purified by Protein A
  • Fp:
    100 µl

Specifications

About this item

Strongly chemotactic for naive (L-selectinhi) CD4 T-cells and for CD8 T-cells and weakly attractive for resting B-cells and memory (L-selectinlo) CD4 T-cells. May play a role in promoting encounters between recirculating T-cells and dendritic cells and in the migration of activated B-cells into the T-zone of secondary lymphoid tissues. Binds to chemokine receptor CCR7. Binds to atypical chemokine receptor ACKR4 and mediates the recruitment of beta-arrestin (ARRB1/2) to ACKR4.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCL19
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat