Anti-RAP2B Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-2330R-A555
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Ras-related protein Rap-2b
- Antigen symbol:RAP2B
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:5912
- Antigen synonyms:Ras related protein RAP2B |Ras related protein RAP 2B|RAP 2B|RAP2B member of RAS oncogene family|Small GTP binding protein.RAP2B_HUMAN|Ras family small GTP binding protein RAP2B
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
RAP2B belongs to a family of RAS-related GTP-binding proteins. RAP proteins share approximately 50% amino acid identity with the classical RAS proteins and have numerous structural features in common. However, at their 61st amino acid the glutamine in RAS proteins is replaced by threonine in RAP proteins. RAP2A interacts with phospholipase C, epsilon 1 (PLCE1).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RAP2B
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat