Order Entry
Norway
ContactUsLinkComponent
Anti-SCP2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anti-SCP2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-2014R-A488
Supplier:  Bioss
Anti-SCP2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-2014R-A488
Supplier:  Bioss

Specifications

  • Antistofftype:
    Primary
  • Antigen-navn:
    Sterol Carrier Protein 2
  • Antigensymbol:
    SCP2
  • Klonalitet:
    Polyclonal
  • Konjugering:
    Alexa Fluor® 488
  • Vert:
    Rabbit
  • ImmunoKjemi:
    Yes
  • Isotype:
    IgG
  • Reaktivitet:
    Human,
    Rat,
    Mouse
  • Kyssabsorpsjon:
    No
  • Skjema:
    liquid
  • Gen-ID:
    6342
  • Antigen synonymer:
    NSL-TP|NLTP|SCP-CHI|SCP-X|SCPX|SCP-2
  • Lagringsbuffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Lagringstemp:
    Store at 4°C for 12 months
  • Konsentrasjon:
    1 μg/μl
  • Frakttemp.:
    4°C
  • Immunogen:
    460-510/547
  • Rensing:
    Purified by Protein A
  • Fp:
    100 µl

Specifications

About this item

Mediates in vitro the transfer of all common phospholipids, cholesterol and gangliosides between membranes. May play a role in regulating steroidogenesis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SCP2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat