Anti-SLC39A7 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
BOSSBS-2391R-HRP
: Bioss
- Antibody type:Primary
- Antigen name:Solute Carrier Family 39 Member A7
- Antigen symbol:SLC39A7
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Gene ID:7922
- Antigen synonyms:HKE4|H2-KE4|D6S115E|Zinc transporter SLC39A7|D6S2244E|RING5|Histidine-rich membrane protein Ke4|KE4|ZIP7
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Zinc is an essential cofactor for more than 50 classes of enzymes and is involved in protein, nucleic acid, carbohydrate, and lipid metabolism, plus the control of gene transcription, growth, development, and differentiation. Zinc cannot passively diffuse across cell membranes and requires specific transporters, such as SLC39A7, to enter the cytosol from both the extracellular environment and from intracellular storage compartments.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: SLC39A7
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat