Anti-SEL1L Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-1891R-A647
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:SEL1L ERAD E3 Ligase Adaptor Subunit
- Antigen symbol:SEL1L
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:6400
- Antigen synonyms:SEL1L1|PRO1063|SEL1-LIKE
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:180-230/794
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
May play a role in Notch signaling (By similarity). May be involved in the endoplasmic reticulum quality control (ERQC) system also called ER-associated degradation (ERAD) involved in ubiquitin-dependent degradation of misfolded endoplasmic reticulum proteins.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SEL1L
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat