Anti-IL7 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-1811R-A488
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 7
- Antigen symbol:IL7
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:16196
- Antigen synonyms:IL-7|IL7|Interluekin-7|Interluekin 7|IL 7|interluekin
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:50-100/154
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Hematopoietic growth factor capable of stimulating the proliferation of lymphoid progenitors. It is important for proliferation during certain stages of B-cell maturation.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: IL7
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat