Anti-GZMB Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-1351R-A350
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Granzyme B
- Antigen symbol:GZMB
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:3002
- Antigen synonyms:CSP-B|Granzyme 2 cytotoxic T-lymphocyte-associated serine esterase 1|CCPI|CTSGL1|CGL1|CGL-1|SECT|CSPB|CTLA1|HLP
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:5-55/247
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
This enzyme is necessary for target cell lysis in cell-mediated immune responses. It cleaves after Asp. Seems to be linked to an activation cascade of caspases (aspartate-specific cysteine proteases) responsible for apoptosis execution. Cleaves caspase-3, -7, -9 and 10 to give rise to active enzymes mediating apoptosis.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: pan Granzyme
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat