Anti-GOLGA6L4 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-13483R-CY7
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Golgin A6 Family like 4
- Antigen symbol:GOLGA6L4
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:6 like 4|golgin subfamily a|Golgi autoantigen|GG6L4_HUMAN.|Putative golgin subfamily A member 6 like protein 4
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Belongs to the GOLGA6 family.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GOLGA6L4
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human