Anti-GLG1 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-13485R-CY5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Golgi Glycoprotein 1
- Antigen symbol:GLG1
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:2734
- Antigen synonyms:AW537898|CFR|MG-160|ESL-1|AI593353|MG160|Selel|CFR-1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Binds fibroblast growth factor and E-selectin (cell-adhesion lectin on endothelial cells mediating the binding of neutrophils).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Golgi Complex
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat