Anti-Glucoamylase Rabbit Polyclonal Antibody (Alexa Fluor® 488)
BOSSBS-13383R-A488
: Bioss
- Antibody type:Primary
- Antigen name:Glucoamylase
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Multiple
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:Glucan 1,4|AMYG_ASPAW|Glucoamylase 3|Glucan 14 alpha glucosidase|Amyloglucosidase|Glucoamylase|Glucan 1 4 alpha glucosidase|Gluc 2|1,4-alpha-D-glucan glucohydrolase|1 4 alpha D glucan glucohydrolase|Gluc 1|Gluc 3|Glucoamylase 2|Glucoamylase 1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Glucoamylase is an important industrial enzyme used in saccharification steps in both starch enzymatic conversion and in alcohol production. The catalytic domain degrades oligosaccharides from the non reducing end, releasing glucose, and the starch domain binds the enzyme to raw starch and to the cell wall.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Glucoamylase
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Other Species