- Antibody type:Primary
- Antigen name:Membrane Bound O-Acyltransferase Domain Containing 4
- Antigen symbol:MBOAT4
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:619373
- Antigen synonyms:FKSG89|GOAT|OACT4
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
MBOAT4 (membrane-bound O-acyltransferase domain-containing protein 4, Ghrelin O-acyltransferase) is a 435 amino acid, multi-pass membrane protein that belongs to the membrane-bound acyltransferase family. MBOAT4 functions as an enzyme that attaches an octanoate fatty acid to serine-3 of ghrelin. Ghrelin is a very small, appetite-stimulating hormone secreted by the food-deprived stomach. MBOAT4 can use a variety of fatty acids as substrates including octanoic acid, decanoic acid and tetradecanoic acid. MBOAT4 expression, consistent with its function, is mainly in the stomach and intestines. Due to its primary function, MBOAT4 is a candidate for obesity and appetite suppression studies.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: MBOAT4
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat