Specifications
- Antibody type:Primary
- Antigen name:GCN1 eIF2 alpha kinase activator homolog
- Antigen symbol:GCN1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:10985
- Antigen synonyms:GCN1L|GCN1L1|PRIC295
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
GCN1L1 is a 2,671 amino acid protein that is ubiquitously expressed and belongs to the GCN1 family. Functioning as a translation activator, GCN1L1 interacts with IMPACT to regulproteinate GCN2 kinase activity. GCN1L1 contains 24 HEAT repeats and is encoded by a gene that maps to human chromosome 12q24.23. Chromosome 12 encodes over 1,100 genes and comprises approximately 4.5% of the human genome. Chromosome 12 is associated with a variety of diseases and afflictions, including hypochondrogenesis, achondrogenesis, Kniest dysplasia, Noonan syndrome and trisomy 12p, which causes facial developmental defects and seizure disorders.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Gcn1l1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat