Specifications
- Antibody type:Primary
- Antigen name:Finkel-Biskis-Reilly murine sarcoma virus (FBR-MuSV) ubiquitously expressed
- Antigen symbol:FAU
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:2197
- Antigen synonyms:Fubi|MNSFbeta|RPS30|FAU1|S30|Fub1|asr1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
The ubiquitin (Ub) pathway involves three sequential enzymatic steps that facilitate the conjugation of Ub and Ub-like molecules to specific protein substrates. The first step requires the ATP-dependent activation of the Ub C-terminus and the assembly of multi-Ub chains by the Ub-activating enzyme known as the E1 component. The Ub chain is then conjugated to the Ub-conjugating enzyme (E2) to generate an intermediate Ub-E2 complex. The Ub-ligase (E3) then catalyzes the transfer of Ub from E2 to the appropriate protein substrate. A wide range of enzymes facilitate in the proteolytic Ub pathway, including monoclonal nonspecific suppressor factor-beta (MNSF-beta), a subunit of MNSF, which is a lymphokine product of a murine T cell hybrid-oma that restricts the production of LPS-induced immunoglobulin secreting cells in an antigen-nonspecific manner. MNSF-beta is a ubiquitin-like fusion protein consisting of the ribosomal protein S30 and a protein that shares 36% sequence identity with ubiquitin. This ubiquitin-like segment (Ubi-L) can be cleaved from MNSF-beta in the cytosol.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: FAU/MNSF-beta
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat