Anti-HLA-E Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
BOSSBS-1279R-FITC
: Bioss
- Antibody type:Primary
- Antigen name:HLA class I histocompatibility antigen, E alpha chain
- Antigen symbol:HLA-E
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Gene ID:3133
- Antigen synonyms:EA2.1|HLA class I histocompatibility antigen|HLA-6.2|lymphocyte antigen|MHC HLA-E alpha-1|E alpha chain|DAMA-277I14.1|DKFZp686P19218|MHC class I antigen E|EA1.2|MHC|alpha chain E|QA1|MHC HLA-E alpha-2.1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:330-358/358
- Purification:Purified by Protein A
- Pk:100 µl
HLA E belongs to the HLA class I heavy chain paralogues. This class I molecule is a heterodimer consisting of a heavy chain and a light chain (beta-2 microglobulin). The heavy chain is anchored in the membrane. HLA E binds a restricted subset of peptides derived from the leader peptides of other class I molecules.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HLA-E
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human