Order Entry
Norway
ContactUsLinkComponent
 
Anti-HLA-E Rabbit Polyclonal Antibody (Alexa Fluor® 647)
  BOSSBS-1279R-A647
 :  Bioss
Anti-HLA-E Rabbit Polyclonal Antibody (Alexa Fluor® 647)
  BOSSBS-1279R-A647
 :  Bioss

 

  • Antistofftype:
    Primary
  • Antigen-navn:
    HLA class I histocompatibility antigen, E alpha chain
  • Antigensymbol:
    HLA-E
  • Klonalitet:
    Polyclonal
  • Konjugering:
    Alexa Fluor® 647
  • Vert:
    Rabbit
  • ImmunoKjemi:
    Yes
  • Isotype:
    IgG
  • Reaktivitet:
    Human
  • Kyssabsorpsjon:
    No
  • Skjema:
    liquid
  • Gen-ID:
    3133
  • Antigen synonymer:
    EA2.1|HLA class I histocompatibility antigen|HLA-6.2|lymphocyte antigen|MHC HLA-E alpha-1|E alpha chain|DAMA-277I14.1|DKFZp686P19218|MHC class I antigen E|EA1.2|MHC|alpha chain E|QA1|MHC HLA-E alpha-2.1
  • Lagringsbuffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Lagringstemp:
    Store at 4°C for 12 months
  • Konsentrasjon:
    1 μg/μl
  • Frakttemp.:
    4°C
  • Immunogen:
    330-358/358
  • Rensing:
    Purified by Protein A
  • Fp:
    100 µl

 

 

HLA E belongs to the HLA class I heavy chain paralogues. This class I molecule is a heterodimer consisting of a heavy chain and a light chain (beta-2 microglobulin). The heavy chain is anchored in the membrane. HLA E binds a restricted subset of peptides derived from the leader peptides of other class I molecules.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HLA-E
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human