Anti-ABTB1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
BOSSBS-12881R-A555
: Bioss
- Antibody type:Primary
- Antigen name:Ankyrin Repeat And BTB (POZ) Domain Containing 1
- Antigen symbol:ABTB1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:80325
- Antigen synonyms:BPOZ|Btb3|EF1ABP|PP2259|MGC20585|elongation factor 1A binding protein|ABTB1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
ABTB1 is a 478 amino acid protein localized to the cytoplasm. ABTB1 contains two ANK repeats and two BTB (POZ) domains. The BTB (POZ) domain is thought to be involved in protein-protein interactions, and may indicate a role of ABTB1 in developmental processes. It has also been suggested that ABTB1 may be a mediator of the PTEN growth-suppressive signaling pathway. ABTB1 is ubiquitously expressed in all fetal tissues, with lower levels of expression found in adult heart. ABTB1 exists as four isoforms produced by alternative splicing.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ABTB1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat