Specifications
- Antibody type:Primary
- Antigen name:1-Acylglycerol-3-Phosphate O-Acyltransferase 5
- Antigen symbol:AGPAT5
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:55326
- Antigen synonyms:LPAATE|1AGPAT5
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Phosphatidic acid and lysophosphatidic acid are phospholipids involved in lipid biosynthesis and signal transduction. LPAAT-epsilon (lysophosphatidic acid acyltransferase epsilon, also designated 1-AGP acyltransferase 5 (AGPAT5)) catalyzes the synthesis of phosphatidic acid from lysophosphatidic acid. LPAAT-epsilon is a membrane-bound protein belonging to the LPAAT family. Members of the LPAAT family have a well-known role in lipid biosynthesis and they may also play a role in tumor progression. LPAAT-epsilon is expressed in a tissue-specific manner in prostate and testis. LPAAT-epsilon is most closely related to AGPAT8, which is highly expressed in heart.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: AGPAT5
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat