Specifications
- Antibody type:Primary
- Antigen name:Cyclin And CBS Domain divalent metal cation transport mediator 3
- Antigen symbol:CNNM3
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:26505
- Antigen synonyms:ACDP3
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Cyclin M3 is a 707 amino acid multi-pass membrane protein that shares weak sequence similarity with cyclin proteins, yet displays no cyclin-like function in vivo. Though ubiquitously expressed, Cyclin M3 is found at highest levels in kidney, brain, spleen and heart. Cyclin M3 is localized to the nucleus where it is likely a metal transporter. Cyclin M3 contains two CBS domains, which appear to bind ligands with an adenosyl group such as AMP, ATP and S-AdoMet and may play a regulatory role in sensitizing proteins to adenosyl-carrying ligands. There are three isoforms of Cyclin M3 that are produced as a result of alternative splicing events.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CNNM3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype:
Reactivity: