Anti-ADGRG6 Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-12027R-CY3
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Adhesion G Protein-Coupled Receptor G6
- Antigen symbol:ADGRG6
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:57211
- Antigen synonyms:GPR126|PS1TP2|DREG|VIGR|APG1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:765-810/1221
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Orphan receptor. May be required for normal differentiation of promyelinating Schwann cells and for normal myelination of axons. Signals probably through G-proteins to transiently elevate cAMP levels (By similarity).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ADGRG6
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat