Specifications
- Antibody type:Primary
- Antigen name:Serine/threonine-protein kinase MAK-V
- Antigen symbol:MAK
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
The HUNK (hormonally upregulated Neu-associated kinase) protein, also designated MAK-V in mouse, has been identified as a novel SNF1-related serine/threonine kinase. The human HUNK gene localizes to chromosome 21q22 and encodes a protein with nucleocytoplasmic distribution and localizes to the centrosome. Overexpression of the HUNK protein associates with approximately 50% of breast carcinomas, and may provide diagnostic-prognostic value as a molecular marker. Serine/threonine-protein kinase SNF1-like kinase 2 (SIK) phosphorylates Ser-794 of IRS1 in insulin-stimulated adipocytes, which may modulate the efficiency of insulin signal transduction. SIK is activated by phosphorylation on Thr-175 by STK11 in complex with STE20-related adapter-α and CAB39.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Serine/threonine-protein kinase MAK-V
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat