Spesifikasjoner
- Antibody type:Primary
- Antigen name:Nucleoside diphosphate kinase D
- Antigen symbol:NME4
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:NDP kinase|mitochondrial|Nucleoside diphosphate kinase|NDK|nm23-H4
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Spesifikasjoner
Om dette produktet
The nm23 gene (Metastasis inhibition factor nm23), a potential suppressor of metastasis, is expressed at a much lower level in highly metastatic cells than in cells with lower metastatic potential. Based on sequence analysis, nm23, also designated nucleoside diphosphate kinase A (NDK A) or Tumor metastatic process-associated protein, appears to be highly related to nucleotide diphosphate kinases (NDP). NDP kinases A and B are identical to two isotypes of human nm23 homologs, nm23-H1 and nm23-H2, respectively. nm23-H2 is also identical in sequence to PuF, a transcription factor that binds to nuclease-hypersensitive elements at positions 142 to 115 of the human c-Myc promoter. nm23-H3 and nm23-H4 are important for the synthesis of nucleoside triphosphates and may play a role in apoptosis induction and hematopoiesis. nm23-H4 localizes to the mitochondrial intermembrane space and is widely expressed, with higher levels detected in prostate, heart, liver, small intestine, and skeletal muscle tissues. Low amounts of nm23-H4 are observed in the brain and in blood leukocytes.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: NME4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat