Specifications
- Antibody type:Primary
- Antigen name:Immunoglobin-like Domain-Containing protein 4
- Antigen symbol:LINGO4
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:LRRN6D|DAAT9248|PRO34002
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Lingo-4 is a 593 amino acid single-pass type I membrane protein that contains eleven LRR (leucine-rich) repeats, one Ig-like C2-type (immunoglobulin-like) domain, one LRRCT domain and one LRRNT domain. The gene that encodes Lingo-4 consists of approximately 5,891 bases and maps to human chromosome 1q21.3. Comprising nearly 8% of the human genome, chromosome 1 spans 260 million base pairs, contains over 3,000 genes and houses a large number of disease-associated genes, including those that are involved in familial adenomatous polyposis, Stickler syndrome, Parkinson’s disease, Gaucher disease, schizophrenia and Usher syndrome.
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200
Type: Primary
Antigen: LINGO4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat