- Antibody type:Primary
- Antigen name:Paired-like Homeobox 2a
- Antigen symbol:PHOX2A
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:PHX2A_HUMAN|ARIX|Paired like aristaless Homeobox 2A|Paired mesoderm homeobox protein 2A|ARIX1 homeodomain protein|MGC52227|CFEOM 2|FEOM2|ARIX 1 homeodomain protein|NCAM 2|PHOX 2A|Arix homeodomain protein|FEOM 2|Phox2|CFEOM2|Aristaless homeobox Drosophila fibrosis of extraocular muscles congenital 2 autosomal recessive|Paired mesoderm homeobox 2a|NCAM2|Pmx2a.|Fibrosis of extraocular muscles congenital 2 autosomal recessive|Aristaless homeobox protein homolog|Aristaless homeobox gene homolog Drosophila|Paired like homeobox 2a|Paired-like homeobox 2A|Pmx 2a|Pmx2|Aristaless homeobox homolog|Phox2a
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Phox2a (also designated Arix1) and Phox2b are closely related, paired-homeodomain transcription factors that are necessary for neuronal differentiation throughout the developing sympathetic, parasympathetic and enteric ganglia. All enteric nervous system cells evolve from the neural crest, and all cells that are undifferentiated initially express Phox2b. The cells that begin to differentiate along a neuronal lineage continue to express Phox2b, and begin to express Phox2a. Phox2b is required for the differentiation of all central and nonperipheral noradrenergic centers in the brain. In contrast, Phox2a controls only the differentiation of the main noradrenergic center of the brain, the locus ceruleus. Both Phox2a and Phox2b are crucial for the regulation of endogenous tyrosine hydroxylase and dopamine-beta hydroxylase, which are transiently expressed in neural crest cells. In addition, Phox2 proteins are sufficient to promote sympathetic neuron generation. The gene which encodes Phox2a maps to human chromosome 11q13.3-q13.4.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PHOX2A
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat