- Antibody type:Primary
- Antigen name:Endosulfine alpha
- Antigen symbol:ENSA
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:ARPP-19e|MGC78563|MGC8394|MGC4319
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Sulfonylureas are a class of drugs commonly used in the management of noninsulin-dependent diabetes mellitus. ENSA/ARPP-19, also known as ARPP-19e, is a 121 amino acid protein that localizes to the cytoplasm and belongs to the endosulfine family. ENSA/ARPP-19 is an endogenous regulator of the β-cell K(ATP) channels and reduces K(ATP) channel currents by inhibiting sulfonylurea from binding to the receptor thereby stimulating insulin secretion by pancreatic β cells. It is suggested that ENSA/ARPP-19 expression is decreased in brains of both Alzheimer's disease (AD) and Down's syndrome patients. Widely expressed with high levels in skeletal muscle and brain and lower levels in pancreas, ENSA/ARPP-19 is phosphorylated by PKA and exists as eight alternatively spliced isoforms. ENSA/ARPP-19 is encoded by a gene located on human chromosome 1q21.3.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ENSA
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat