Order Entry
Norway
ContactUsLinkComponent
Anti-PS1 + PS2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anti-PS1 + PS2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-11681R-A647
Supplier:  Bioss
Anti-PS1 + PS2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-11681R-A647
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Presenilin 1 + 2
  • Antigen symbol:
    PS1 + PS2
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 647
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Antigen synonyms:
    PS2|AD 3|FAD|AD4|Alzheimer disease 3|Presenilin 2|Presenilin 1|PS-2|PS1|PSEN2|Presenilin-1|PSN1_HUMAN|PSEN1|Alzheimer disease 4|PSN2_HUMAN.|STM2|PS-1|Presenilin1|AD5|Presenilin-2
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Alzheimer's disease (AD) patients with an inherited form of the disease carry mutations in the presenilin proteins (PSEN1; PSEN2) or the amyloid precursor protein (APP). These disease-linked mutations result in increased production of the longer form of amyloid-beta (main component of amyloid deposits found in AD brains). Presenilins are postulated to regulate APP processing through their effects on gamma-secretase, an endoprotease complex that catalyzes the intramembrane cleavage of integral membrane proteins such as Notch receptors and APP.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PS1 + PS2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat