Spesifikasjoner
- Antibody type:Primary
- Antigen name:FERM, RhoGEF and Pleckstrin Domain Protein 2
- Antigen symbol:FARP2
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:FARP2|FERM|PLEKHC3|RhoGEF and pleckstrin domain-containing protein 2|KIAA0793|FIR|PH domain-containing family C member 3|Pleckstrin homology domain-containing family C member 3|FERM domain including RhoGEF
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Spesifikasjoner
Om dette produktet
FARP2 is a 1,545 amino acid protein that contains one FERM domain, one DH domain and two PH domains. It exists as two alternatively spliced isoforms that are abundantly expressed in brain, lung, and testis as well as in embryonic hippocampal and cortical neurons. FARP2 functions as a Rho-guanine nucleotide exchange factor that activates RAC1 and is thought to regulate neurite remodeling of embryonic neurons. Sema3A binding to neuropilin-1 induces the dissociation of FARP2 from plexin-A1, thereby activating FARP2's Rac GEF activity which is critical for repulsion of outgrowing axons and suppression of neuronal adhesion. Downregulation of the FARP2 gene has been implicated in autism.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: FARP2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat