Order Entry
Norway
ContactUsLinkComponent
Anti-PTPZ Rabbit Polyclonal Antibody (Cy7®)
Anti-PTPZ Rabbit Polyclonal Antibody (Cy7®)
  BOSSBS-11327R-CY7
 :  Bioss
Anti-PTPZ Rabbit Polyclonal Antibody (Cy7®)
  BOSSBS-11327R-CY7
 :  Bioss

 

  • Antistofftype:
    Primary
  • Antigen-navn:
    Protein Tyrosine Phosphatase zeta
  • Antigensymbol:
    PTPZ
  • Klonalitet:
    Polyclonal
  • Konjugering:
    Cy7®
  • Flowcytometri:
    Yes
  • Vert:
    Rabbit
  • ImmunoKjemi:
    Yes
  • Isotype:
    IgG
  • Reaktivitet:
    Human,
    Rat,
    Mouse
  • Kyssabsorpsjon:
    No
  • Skjema:
    liquid
  • Gen-ID:
    5803
  • Lagringsbuffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Lagringstemp:
    Store at 4°C for 12 months
  • Konsentrasjon:
    1 μg/μl
  • Frakttemp.:
    4°C
  • Immunogen:
    460-500/2315
  • Rensing:
    Purified by Protein A
  • Fp:
    100 µl

 

 

Protein tyrosine phosphatase that negatively regulates oligodendrocyte precursor proliferation in the embryonic spinal cord. Required for normal differentiation of the precursor cells into mature, fully myelinating oligodendrocytes. May play a role in protecting oligondendrocytes against apoptosis. May play a role in the establishment of contextual memory, probably via the dephosphorylation of proteins that are part of important signaling cascades (By similarity).

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: PTP zeta
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat