Specifications
- Antibody type:Primary
- Antigen name:Phosphate cytidylyltransferase 1, Choline, alpha
- Antigen symbol:PCYT1A
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:CT|CCTA|PCYT1|CTPCT|CTA|SMDCRD
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Increase in fetal surfactant synthesis and lung maturity is caused by the glucocorticoidal induction of enzymes required for phosphatidylcholine synthesis towards the end of gestation (1). The regulation of gestational age-dependent induction of phosphatidylcholine synthesis by glucocorticoids is still unclear (1). The rate-controlling enzyme in the phosphatidylcholine biosynthetic pathway is CTP-phosphocholine cytidylyltransferase A (CCT A) (2–4). In cultured eukaryotic cells, this enzyme is essential for survival (3). The alpha isoform is located in the nucleus and is regulated by reversible phosphorylation and membrane association (3). There is significant identity between the alpha-helical membrane-binding domains of CCT A and soybean oleosin (2). Expressed CCT A has lipid-dependent cytidylyltransferase activity (5). The gene which encodes CCT A maps to human chromosome 3q (4).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PCYT1A
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat