Anti-CD86 Rabbit Polyclonal Antibody (Cy7®)
BOSSBS-11250R-CY7
: Bioss
- Antibody type:Primary
- Antigen name:CD86 molecule
- Antigen symbol:CD86
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:942
- Antigen synonyms:CLS1|B70|B7.2|Ly-58|Cd28l2|ETC-1|MB7|TS/A-2|B7-2|B7|MB7-2|Ly58
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Receptor involved in the costimulatory signal essential for T-lymphocyte proliferation and interleukin-2 production, by binding CD28 or CTLA-4. May play a critical role in the early events of T-cell activation and costimulation of naive T-cells, such as deciding between immunity and anergy that is made by T-cells within 24 hours after activation. Isoform 2 interferes with the formation of CD86 clusters, and thus acts as a negative regulator of T-cell activation.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CD86
Clonality: Polyclonal
Clone: 0
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: