Anti-MDGA2 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-11092R-CY7
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:MAM Domain-Containing Glycosylphosphatidylinositol Anchor protein 2
- Antigen symbol:MDGA2
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:161357
- Antigen synonyms:MAM domain-containing glycosylphosphatidylinositol anchor protein 2|MAM domain-containing protein 1|c14_5286|MAMDC1|UNQ8188/PRO23197|MDGA2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:780-830/956
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
May be involved in cell-cell interactions.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: MDGA2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat